accessibility menu, dialog, popup

UserName

CH3OOO

CAS RN 55418-52-5

IUPAC Name: 4-(2H-1,3-benzodioxol-5-yl)butan-2-one
Synonyms: piperonyl acetone methylenedioxyphenyl butanone
Molecular Weight (g/mol): 192.21
Molecular Formula: C11H12O3
InChi Key: TZJLGGWGVLADDN-UHFFFAOYSA-N
SMILES: CC(=O)CCC1=CC=C2OCOC2=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)