CAS RN 554407-47-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl 4-[(Pyridine-3-carbonylamino)methyl]benzoate, TRC
CAS: 554407-47-5 Molecular Formula: C15H14N2O3 Molecular Weight (g/mol): 270.28 SMILES: COC(=O)c1ccc(CNC(=O)c2cccnc2)cc1