CAS RN 55534-89-9
IUPAC Name:
bis(5,5,5-trifluoro-4-hydroxypent-3-en-2-one) dihydrate nickel
Synonyms:
bis(5,5,5-trifluoro-4-hydroxypent-3-en-2-one) dihydrate nickel
Molecular Weight (g/mol):
402.9
Molecular Formula:
C10H14F6NiO6
InChi Key:
ZKWNTTFKFAZZPW-UHFFFAOYSA-N
SMILES:
O.O.[Ni].CC(=O)C=C(O)C(F)(F)F.CC(=O)C=C(O)C(F)(F)F
1
–
1
of
1
results
1
Nickel(II) 1,1,1-trifluoro-2,4-pentanedionate dihydrate, Thermo Scientific™
CAS: 55534-89-9 Molecular Formula: C10H12F6NiO6 Molecular Weight (g/mol): 400.88 MDL Number: MFCD00150656 InChI Key: TXAVMKXZMQAXHU-UDVCPWNYSA-L Synonym: nickel trifluoroacetylacetonate dihydrate IUPAC Name: nickel(2+) bis((2Z)-5,5,5-trifluoro-4-oxopent-2-en-2-olate) dihydrate SMILES: O.O.[Ni++].C\C([O-])=C\C(=O)C(F)(F)F.C\C([O-])=C\C(=O)C(F)(F)F