accessibility menu, dialog, popup

UserName

CAS RN 5591-70-8

IUPAC Name: 4-phenyl-1H-pyrazol-5-amine
Synonyms: 4-phenyl-2H-pyrazol-3-amine
Molecular Weight (g/mol): 159.19
Molecular Formula: C9H9N3
InChi Key: QEHKQNYBBLCFIJ-UHFFFAOYSA-N
SMILES: NC1=C(C=NN1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results