accessibility menu, dialog, popup

UserName

SOOClN

CAS RN 56106-01-5

IUPAC Name: (3-cyanophenyl)methanesulfonyl chloride
Synonyms: (3-cyanophenyl)methanesulfonyl chloride
Molecular Weight (g/mol): 215.65
Molecular Formula: C8H6ClNO2S
InChi Key: YNWYDZBBHOWDAE-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)CC1=CC=CC(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results