accessibility menu, dialog, popup

UserName

OOO

CAS RN 5617-74-3

IUPAC Name: 3-oxabicyclo[3.1.0]hexane-2,4-dione
Synonyms: 3-oxabicyclo[3.1.0]hexane-2,4-dione
Molecular Weight (g/mol): 112.08
Molecular Formula: C5H4O3
InChi Key: ZRMYHUFDVLRYPN-UHFFFAOYNA-N
SMILES: O=C1OC(=O)C2CC12
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)