accessibility menu, dialog, popup

UserName

CAS RN 5623-90-5

OOO

CAS RN 5623-90-5

IUPAC Name: 2-oxaspiro[4.4]nonane-1,3-dione
Synonyms: 2-oxaspiro[4.4]nonane-1,3-dione
Molecular Weight (g/mol): 154.17
Molecular Formula: C8H10O3
InChi Key: HOKKSYPZFBKQAZ-UHFFFAOYSA-N
SMILES: O=C1CC2(CCCC2)C(=O)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results