accessibility menu, dialog, popup

UserName

H3CBrBrO

CAS RN 563-76-8

IUPAC Name: 2-bromopropanoyl bromide
Synonyms: 2-bromopropanoyl bromide
Molecular Weight (g/mol): 215.87
Molecular Formula: C3H4Br2O
InChi Key: ILLHORFDXDLILE-UHFFFAOYNA-N
SMILES: CC(Br)C(Br)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results