accessibility menu, dialog, popup

UserName

NaHO(R)O(R)OPOHOHO(R)OH(S)HO(S)HO

CAS RN 56401-20-8

IUPAC Name: sodium {[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}phosphonic acid
Synonyms: sodium α-D-glucose 1-phosphate
Molecular Weight (g/mol): 283.12
Molecular Formula: C6H13NaO9P
InChi Key: YSLVOZPKBHMBRV-WYRLRVFGSA-N
SMILES: [Na+].OC[C@H]1O[C@H](OP(O)(O)=O)[C@H](O)[C@@H](O)[C@@H]1O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)