accessibility menu, dialog, popup

UserName

CAS RN 565-53-7

H3COOFBrBr

CAS RN 565-53-7

IUPAC Name: ethyl 2,2-dibromo-2-fluoroacetate
Synonyms: ethyl 2,2-dibromo-2-fluoroacetate
Molecular Weight (g/mol): 263.89
Molecular Formula: C4H5Br2FO2
InChi Key: COYRDMUQLSIAEX-UHFFFAOYSA-N
SMILES: CCOC(=O)C(F)(Br)Br
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results