CAS RN 565-74-2
CAS RN 565-74-2
IUPAC Name:
2-bromo-3-methylbutanoic acid
Synonyms:
2-bromo-3-methyl-butanoic acid
Molecular Weight (g/mol):
181.03
Molecular Formula:
C5H9BrO2
InChi Key:
UEBARDWJXBGYEJ-UHFFFAOYNA-N
SMILES:
CC(C)C(Br)C(O)=O
1
–
2
of
2
results
1
2-Bromoisovaleric acid, 96%
CAS: 565-74-2 Molecular Formula: C5H9BrO2 Molecular Weight (g/mol): 181.029 MDL Number: MFCD00004210 InChI Key: UEBARDWJXBGYEJ-UHFFFAOYSA-N Synonym: 2-bromo-3-methylbutyric acid,2-bromoisovaleric acid,butanoic acid, 2-bromo-3-methyl,alpha-bromoisovaleric acid,butyric acid, 2-bromo-3-methyl,2-bromo-3-methyl-butanoic acid,.alpha.-bromoisovaleric acid,+/--2-bromo-3-methylbutyric acid,bromisovaleriansaure,acmc-209wpb PubChem CID: 11268 IUPAC Name: 2-bromo-3-methylbutanoic acid SMILES: CC(C)C(C(=O)O)Br