accessibility menu, dialog, popup

UserName

CAS RN 565-75-3

CH3H3CCH3CH3H3C

CAS RN 565-75-3

IUPAC Name: 2,3,4-trimethylpentane
Synonyms: 2,3,4-trimethylpentane
Molecular Weight (g/mol): 114.23
Molecular Formula: C8H18
InChi Key: RLPGDEORIPLBNF-UHFFFAOYSA-N
SMILES: CC(C)C(C)C(C)C
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)