accessibility menu, dialog, popup

UserName

H3CONCO

CAS RN 56651-60-6

IUPAC Name: 1-(isocyanatomethyl)-4-methoxybenzene
Synonyms: 1-(isocyanatomethyl)-4-methoxybenzene
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: QRBHVARIMDDOOV-UHFFFAOYSA-N
SMILES: COC1=CC=C(CN=C=O)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results