CAS RN 56742-45-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Methyl Metribuzin, TRC
CAS: 56742-45-1 Molecular Formula: C9 H16 N4 O S Molecular Weight (g/mol): 228.31 Synonym: 6-(1,1-Dimethylethyl)-4-(methylamino)-3-(methylthio)-1,2,4-triazin-5(4H)-one,6-tert-Butyl-4-(methylamino)-3-(methylthio)-1,2,4-triazin-5(4H)-one IUPAC Name: 6-tert-butyl-4-(methylamino)-3-methylsulfanyl-1,2,4-triazin-5-one SMILES: CNN1C(=NN=C(C1=O)C(C)(C)C)SC