accessibility menu, dialog, popup

UserName

CH3OOOOOCH3

CAS RN 5676-71-1

IUPAC Name: bis(4-methoxyphenyl) carbonate
Synonyms: bis(4-methoxyphenyl) carbonate
Molecular Weight (g/mol): 274.27
Molecular Formula: C15H14O5
InChi Key: PVBTWSPZTNYNHG-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC(=O)OC2=CC=C(OC)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results