CAS RN 57336-69-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(E)-Cyano(methoxyimino)-acetic Acid, TRC
CAS: 57336-69-3 Molecular Formula: C4 H4 N2 O3 Molecular Weight (g/mol): 128.09 Synonym: (E)-2-Cyano-2-(methoxyimino)-acetic Acid IUPAC Name: (2E)-2-cyano-2-methoxyimino-acetic acid SMILES: CO\N=C(/C#N)\C(=O)O