accessibility menu, dialog, popup

UserName

CH3NNH3COHO

CAS RN 5744-59-2

IUPAC Name: 1,5-dimethyl-1H-pyrazole-3-carboxylic acid
Synonyms: 1,5-dimethylpyrazole-3-carboxylic acid
Molecular Weight (g/mol): 140.14
Molecular Formula: C6H8N2O2
InChi Key: PXRXGHUTKHXUGF-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1C)C(O)=O
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results