accessibility menu, dialog, popup

UserName

ClClNNN

CAS RN 57489-77-7

IUPAC Name: 5,7-dichloropyrazolo[1,5-a]pyrimidine
Synonyms: 5,7-dichloropyrazolo[1,5-a]pyrimidine
Molecular Weight (g/mol): 188.01
Molecular Formula: C6H3Cl2N3
InChi Key: JMTFWCYVZOFHLR-UHFFFAOYSA-N
SMILES: ClC1=NC2=CC=NN2C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results