CAS RN 57497-39-9
IUPAC Name:
hydrogen N-tert-butylhydroxylamine chloride
Synonyms:
hydrogen N-tert-butylhydroxylamine chloride
Molecular Weight (g/mol):
125.6
Molecular Formula:
C4H12ClNO
InChi Key:
DCSATTBHEMKGIP-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CC(C)(C)NO