accessibility menu, dialog, popup

UserName

CAS RN 5751-83-7

IUPAC Name: ethyl 5-bromothiophene-2-carboxylate
Synonyms: ethyl 5-bromothiophene-2-carboxylate
Molecular Weight (g/mol): 235.1
Molecular Formula: C7H7BrO2S
InChi Key: PZNHMXAOMDQLLE-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(Br)S1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)