accessibility menu, dialog, popup

UserName

ONONHClN

CAS RN 57531-37-0

IUPAC Name: 2-chloro-5-nitro-1H-imidazole
Synonyms: 2-chloro-4-nitro-3H-imidazole
Molecular Weight (g/mol): 147.52
Molecular Formula: C3H2ClN3O2
InChi Key: BOJZBRDIZUHTCE-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CN=C(Cl)N1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)