CAS RN 57564-91-7
IUPAC Name:
(2S)-2-amino-4-{[(1R)-1-[(carboxymethyl)carbamoyl]-2-(nitrososulfanyl)ethyl]carbamoyl}butanoic acid
Synonyms:
nitrosoglutathione s-nitroso-gsh GSNO SNOG glutathione thionitrite
Molecular Weight (g/mol):
336.32
Molecular Formula:
C10H16N4O7S
InChi Key:
HYHSBSXUHZOYLX-WDSKDSINSA-N
SMILES:
N[C@@H](CCC(=O)N[C@@H](CSN=O)C(=O)NCC(O)=O)C(O)=O
1
–
2
of
2
results
1
S-Nitroso-L-glutathione (>90%), TRC
CAS: 57564-91-7 Synonym: (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-3-nitrososulfanyl-1-oxopropan-2-yl]amino]-5-oxopentanoic acid
2
Tocris Bioscience™ SNOG
CAS: 57564-91-7 Molecular Formula: C10H16N4O7S