CAS RN 57678-01-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Mono[2-(perfluorohexyl)ethyl] phosphate, TRC
CAS: 57678-01-0 Molecular Formula: C8 H6 F13 O4 P Molecular Weight (g/mol): 444.08 Synonym: 6:2 monoPAP,3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-(dihydrogen phosphate)-1-octanol IUPAC Name: 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl dihydrogen phosphate SMILES: OP(=O)(O)OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F