CAS RN 57678-03-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Mono[2-(perfluorooctyl)ethyl] phosphate, TRC
CAS: 57678-03-2 Molecular Formula: C10 H6 F17 O4 P Molecular Weight (g/mol): 544.1 Synonym: 1H,1H,2H,2H-Perfluorodecanephosphate,Mono[2-(perfluorooctyl)ethyl] Phosphate,1-Decanol, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluoro-, dihydrogen phosphate (9CI),Mono[2-(perfluorooctyl)ethyl] phosphate,1-Decanol, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluoro-, 1-(dihydrogen phosphate) IUPAC Name: 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl dihydrogen phosphate SMILES: OP(=O)(O)OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F