accessibility menu, dialog, popup

UserName

CAS RN 57709-61-2

IUPAC Name: 1,10-phenanthroline-2,9-dicarboxylate
Synonyms: 1,10-phenanthroline-2,9-dicarboxylate
Molecular Weight (g/mol): 266.21
Molecular Formula: C14H6N2O4
InChi Key: FXSVCROWUPWXBP-UHFFFAOYSA-L
SMILES: [O-]C(=O)C1=CC=C2C=CC3=CC=C(N=C3C2=N1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results