accessibility menu, dialog, popup

UserName

NH2HNN

CAS RN 5805-39-0

IUPAC Name: 2-(1H-1,3-benzodiazol-2-yl)aniline
Synonyms: 2-(1H-1,3-benzodiazol-2-yl)aniline
Molecular Weight (g/mol): 209.25
Molecular Formula: C13H11N3
InChi Key: YWNXHTNWOQHFRL-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1C1=NC2=CC=CC=C2N1
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results