accessibility menu, dialog, popup

UserName

OH(R)OO

CAS RN 58081-05-3

IUPAC Name: (4R)-4-hydroxyoxolan-2-one
Synonyms: (4R)-4-hydroxyoxolan-2-one
Molecular Weight (g/mol): 102.09
Molecular Formula: C4H6O3
InChi Key: FUDDLSHBRSNCBV-GSVOUGTGSA-N
SMILES: O[C@H]1COC(=O)C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results