accessibility menu, dialog, popup

UserName

H2NOO

CAS RN 5813-90-1

IUPAC Name: 9H-xanthene-9-carboxamide
Synonyms: 9H-xanthene-9-carboxamide
Molecular Weight (g/mol): 225.25
Molecular Formula: C14H11NO2
InChi Key: WEDLKOCAEPRSPJ-UHFFFAOYSA-N
SMILES: NC(=O)C1C2=CC=CC=C2OC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results