CAS RN 5818-06-4
IUPAC Name:
3-hydroxybenzohydrazide
Synonyms:
benzoylhydrazine, M-hydroxy
Molecular Weight (g/mol):
152.15
Molecular Formula:
C7H8N2O2
InChi Key:
OACGSLLKFCMXSX-UHFFFAOYSA-N
SMILES:
NNC(=O)C1=CC=CC(O)=C1
1
–
1
of
1
results
1
3-Hydroxybenzhydrazide, 98+%
CAS: 5818-06-4 MDL Number: MFCD00014759 ChEBI: CHEBI:39411