accessibility menu, dialog, popup

UserName

CAS RN 5825-71-8

CAS RN 5825-71-8

IUPAC Name: 2,3-dimethyl pyrazolo[1,5-a]pyridine-2,3-dicarboxylate
Synonyms: 2,3-dimethyl pyrazolo[1,5-a]pyridine-2,3-dicarboxylate
Molecular Weight (g/mol): 234.21
Molecular Formula: C11H10N2O4
InChi Key: NZJIUQJPKXRRHB-UHFFFAOYSA-N
SMILES: COC(=O)C1=NN2C=CC=CC2=C1C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results