accessibility menu, dialog, popup

UserName

H2NSNHOO

CAS RN 58327-75-6

IUPAC Name: 3-aminothieno[2,3-b]pyridine-2-carboxylic acid
Synonyms: 3-aminothieno[2,3-b]pyridine-2-carboxylic acid
Molecular Weight (g/mol): 194.21
Molecular Formula: C8H6N2O2S
InChi Key: HGKGNTBBCCWIRD-UHFFFAOYSA-N
SMILES: NC1=C(SC2=NC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results