CAS RN 58403-03-5
IUPAC Name:
hydrogen 2-(2-chlorophenoxy)ethanimidamide chloride
Synonyms:
hydrogen 2-(2-chlorophenoxy)ethanimidamide chloride
Molecular Weight (g/mol):
221.08
Molecular Formula:
C8H10Cl2N2O
InChi Key:
WPWKAEFYVGWTAE-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].NC(=N)COC1=CC=CC=C1Cl
1
–
1
of
1
results
1
2-(2-chlorophenoxy)ethanimidamide hydrochloride, 97%, Thermo Scientific™
CAS: 58403-03-5 Molecular Formula: C8H10Cl2N2O Molecular Weight (g/mol): 221.08 MDL Number: MFCD00173805 InChI Key: WPWKAEFYVGWTAE-UHFFFAOYSA-N Synonym: 2-2-chlorophenoxy acetamidine hydrochloride,2-2-chlorophenoxy ethanimidamide hydrochloride,2-2-chlorophenoxy acetimidamide hydrochloride,2-2-chloro-phenoxy-acetamidine hydrochloride,2-2-chlorophenoxy ethanimidamide monohydrochloride,2-2-chlorophenoxy acetamide hydrochloride,ethanimidamide, 2-2-chlorophenoxy-, monohydrochloride,2-2-chlorophenoxy ethanamidine, chloride,2-2-chlorophenoxy acetamidine hcl,2-chlorophenoxy ethanimidamide hydrochloride PubChem CID: 2745914 SMILES: [H+].[Cl-].NC(=N)COC1=CC=CC=C1Cl