accessibility menu, dialog, popup

UserName

CH3H2NHO

CAS RN 5856-63-3

IUPAC Name: 2-aminobutan-1-ol
Synonyms: 2-amino-1-butanol
Molecular Weight (g/mol): 89.14
Molecular Formula: C4H11NO
InChi Key: JCBPETKZIGVZRE-UHFFFAOYNA-N
SMILES: CCC(N)CO
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)