accessibility menu, dialog, popup

UserName

CAS RN 58789-53-0

CH3NNH3CHO

CAS RN 58789-53-0

IUPAC Name: (3,5-dimethyl-1-phenyl-1H-pyrazol-4-yl)methanol
Synonyms: (3,5-dimethyl-1-phenylpyrazol-4-yl)methanol
Molecular Weight (g/mol): 202.26
Molecular Formula: C12H14N2O
InChi Key: JMWWSNPRAOYPSF-UHFFFAOYSA-N
SMILES: CC1=NN(C(C)=C1CO)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results