accessibility menu, dialog, popup

UserName

CAS RN 588-68-1

NN(E)(E)

CAS RN 588-68-1

IUPAC Name: (E,E)-bis(phenylmethylidene)hydrazine
Synonyms: benzalazine
Molecular Weight (g/mol): 208.26
Molecular Formula: C14H12N2
InChi Key: CWLGEPSKQDNHIO-JOBJLJCHSA-N
SMILES: C(=N/N=C/C1=CC=CC=C1)\C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)