accessibility menu, dialog, popup

UserName

ONOON

CAS RN 59-82-5

IUPAC Name: 5-nitrofuran-2-carbonitrile
Synonyms: 5-nitrofuran-2-carbonitrile
Molecular Weight (g/mol): 138.08
Molecular Formula: C5H2N2O3
InChi Key: DUJNJLFQOODDNJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(O1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results