accessibility menu, dialog, popup

UserName

CAS RN 593-71-5

ClI

CAS RN 593-71-5

IUPAC Name: chloro(iodo)methane
Synonyms: chloroiodomethane
Molecular Weight (g/mol): 176.38
Molecular Formula: CH2ClI
InChi Key: PJGJQVRXEUVAFT-UHFFFAOYSA-N
SMILES: ClCI
Boiling Point 
  • (4)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (4)
  • (4)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)