CAS RN 594-31-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
425.99
Molecular Formula:
C18H17Cl2Sb
InChi Key:
WSTBPJVOVGALEQ-UHFFFAOYSA-N
SMILES:
Cl.Cl.C1=CC=C(C=C1)[Sb](C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Triphenylantimony(V) dichloride, 99%, Thermo Scientific Chemicals
CAS: 594-31-0 Molecular Formula: C18H15Cl2Sb Molecular Weight (g/mol): 423.978 MDL Number: MFCD00013598 InChI Key: PDGPVQHGCLPCES-UHFFFAOYSA-L Synonym: triphenylantimony dichloride,antimony, dichlorotriphenyl,dichlorotriphenylantimony,dichlorotriphenylstibine,antimony, triphenyldichloride,triphenylstibine dichloride,dichlorid trifenylantimonicny,dichlorid trifenylantimonicny czech,stibine, triphenyl-, dichloride 6ci,acmc-1ao41 PubChem CID: 11662 IUPAC Name: dichloro(triphenyl)-$l^{5}-stibane SMILES: C1=CC=C(C=C1)[Sb](C2=CC=CC=C2)(C3=CC=CC=C3)(Cl)Cl