accessibility menu, dialog, popup

UserName

H3CCH3CH3H3N

CAS RN 594-39-8

IUPAC Name: 2-methylbutan-2-aminium
Synonyms: 2-methylbutan-2-aminium
Molecular Weight (g/mol): 88.17
Molecular Formula: C5H14N
InChi Key: GELMWIVBBPAMIO-UHFFFAOYSA-O
SMILES: CCC(C)(C)[NH3+]
Boiling Point 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)