accessibility menu, dialog, popup

UserName

CAS RN 594-44-5

IUPAC Name: ethanesulfonyl chloride
Synonyms: ethanesulfonyl chloride
Molecular Weight (g/mol): 128.57
Molecular Formula: C2H5ClO2S
InChi Key: FRYHCSODNHYDPU-UHFFFAOYSA-N
SMILES: CCS(Cl)(=O)=O
Boiling Point 
  • (3)
  • (5)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)