accessibility menu, dialog, popup

UserName

HOOHOOHHOOOHHO

CAS RN 5950-69-6

IUPAC Name: 2,2',3,3,3',3'-hexahydroxy-1H,1'H,2H,2'H,3H,3'H-[2,2'-biindene]-1,1'-dione
Synonyms: 2,2',3,3,3',3'-hexahydroxy-[2,2'-biindene]-1,1'-dione
Molecular Weight (g/mol): 358.3
Molecular Formula: C18H14O8
InChi Key: QHVADKNWNMILPQ-UHFFFAOYNA-N
SMILES: OC1(O)C2=CC=CC=C2C(=O)C1(O)C1(O)C(=O)C2=CC=CC=C2C1(O)O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)