accessibility menu, dialog, popup

UserName

CH3NNOO

CAS RN 5952-92-1

IUPAC Name: 1-methyl-1H-pyrazole-4-carboxylate
Synonyms: 1-methylpyrazole-4-carboxylate
Molecular Weight (g/mol): 125.11
Molecular Formula: C5H5N2O2
InChi Key: UPPPWUOZCSMDTR-UHFFFAOYSA-M
SMILES: CN1C=C(C=N1)C([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results