accessibility menu, dialog, popup

UserName

CAS RN 596-03-2

IUPAC Name: 4',5'-dibromo-3',6'-dihydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Synonyms: 4',5'-dibromofluorescein
Molecular Weight (g/mol): 490.1
Molecular Formula: C20H10Br2O5
InChi Key: ZDTNHRWWURISAA-UHFFFAOYSA-N
SMILES: OC1=CC=C2C(OC3=C(Br)C(O)=CC=C3C22OC(=O)C3=CC=CC=C23)=C1Br
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)