accessibility menu, dialog, popup

UserName

CH3H3COOCH3OOH3C

CAS RN 596-75-8

IUPAC Name: 1,3-diethyl 2,2-dibutylpropanedioate
Synonyms: 1,3-diethyl 2,2-dibutylpropanedioate
Molecular Weight (g/mol): 272.39
Molecular Formula: C15H28O4
InChi Key: WHKKUUPZLWUOIW-UHFFFAOYSA-N
SMILES: CCCCC(CCCC)(C(=O)OCC)C(=O)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)