accessibility menu, dialog, popup

UserName

H3COON(E/Z)

CAS RN 59607-99-7

IUPAC Name: ethyl 3-(pyridin-3-yl)prop-2-enoate
Synonyms: ethyl 3-(pyridin-3-yl)prop-2-enoate
Molecular Weight (g/mol): 177.2
Molecular Formula: C10H11NO2
InChi Key: PIEQSBWGIODYPE-UHFFFAOYSA-N
SMILES: CCOC(=O)C=CC1=CN=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results