CAS RN 5968-90-1
IUPAC Name:
9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-2,3,6,9-tetrahydro-1H-purine-2,6-dione
Synonyms:
xanthosine xanthine riboside 9-delta-ribofuranosylxanthine β-D-ribofuranoside, xanthine-9 xanthine 9-β-D-ribofuranoside xanthine-9b-D-ribofuranoside
Molecular Weight (g/mol):
284.23
Molecular Formula:
C10H12N4O6
InChi Key:
UBORTCNDUKBEOP-UUOKFMHZSA-N
SMILES:
OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C1NC(=O)NC2=O
1
–
2
of
2
results
1
Thermo Scientific Chemicals Xanthosine dihydrate, 99%
CAS: 5968-90-1 Molecular Formula: C10H12N4O6·2H2O Molecular Weight (g/mol): 320.26 InChI Key: AQQAMLPKPXTPOL-GWTDSMLYSA-N Synonym: xanthosine dihydrate,unii-1u62i480sp,9-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purine-2,6-diol dihydrate,xanthosine, dihydrate,xanthosine dihydrate mi,xanthosine,dihydrate,9-beta-d-ribofuranosyl xanthine dihydrate,9-,a-d-ribofuranosylxanthine,xanthosine, dihydrate 8ci,9ci PubChem CID: 91886582 IUPAC Name: 9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purine-2,6-dione;hydrate SMILES: C1=NC2=C(N1C3C(C(C(O3)CO)O)O)NC(=O)NC2=O.O