CAS RN 597-49-9
IUPAC Name:
3-ethylpentan-3-ol
Synonyms:
3-ethyl-3-pentanol
Molecular Weight (g/mol):
116.2
Molecular Formula:
C7H16O
InChi Key:
XKIRHOWVQWCYBT-UHFFFAOYSA-N
SMILES:
CCC(O)(CC)CC
1
–
1
of
1
results
1
3-Ethyl-3-pentanol, 97%
CAS: 597-49-9 Molecular Formula: C7H16O Molecular Weight (g/mol): 116.20 MDL Number: MFCD00004484 InChI Key: XKIRHOWVQWCYBT-UHFFFAOYSA-N Synonym: 3-ethyl-3-pentanol,triethylcarbinol,3-pentanol, 3-ethyl,triethylmethanol,triethylmethyl alcohol,tert-heptanol,3-aethyl-pentanol-3,3-aethyl-pentanol-3 german,3-ethyl-3-hydroxypentane,ethyl-3 pentanol-3 PubChem CID: 11702 IUPAC Name: 3-ethylpentan-3-ol SMILES: CCC(O)(CC)CC