accessibility menu, dialog, popup

UserName

CAS RN 5978-70-1

H3C(R)CH3OH

CAS RN 5978-70-1

IUPAC Name: (2R)-octan-2-ol
Synonyms: (R)-2-octanol 2-octanol, (theta)- (theta)-2-octanol l-octan-2-ol
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: SJWFXCIHNDVPSH-MRVPVSSYSA-N
SMILES: CCCCCC[C@@H](C)O
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)