accessibility menu, dialog, popup

UserName

PPClClPd

CAS RN 59831-02-6

IUPAC Name: palladium(2+) [3-(diphenylphosphaniumyl)propyl]diphenylphosphanium dichloride
Synonyms: palladium(2+) [3-(diphenylphosphaniumyl)propyl]diphenylphosphanium dichloride
Molecular Weight (g/mol): 591.79
Molecular Formula: C27H28Cl2P2Pd
InChi Key: LDFBXJODFADZBN-UHFFFAOYSA-N
SMILES: [Cl-].[Cl-].[Pd++].C(C[PH+](C1=CC=CC=C1)C1=CC=CC=C1)C[PH+](C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)