accessibility menu, dialog, popup

UserName

CAS RN 59843-77-5

IUPAC Name: 5-(3-nitrophenyl)-1H-pyrazole
Synonyms: 3-(3-nitrophenyl)-2H-pyrazole
Molecular Weight (g/mol): 189.17
Molecular Formula: C9H7N3O2
InChi Key: RUWXJVIBNZSEQD-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(=C1)C1=CC=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results